Merck
Histone deacetylase 7 controls endothelial cell growth through modulation of beta-catenin.
Circulation research (2010-03-13)
Andriana Margariti, Anna Zampetaki, Qingzhong Xiao, Boda Zhou, Eirini Karamariti, Daniel Martin, Xiaoke Yin, Manuel Mayr, Hongling Li, Zhongyi Zhang, Elena De Falco, Yanhua Hu, Gillian Cockerill, Qingbo Xu, Lingfang Zeng
ZUSAMMENFASSUNG
Histone deacetylase (HDAC)7 is expressed in the early stages of embryonic development and may play a role in endothelial function. This study aimed to investigate the role of HDAC7 in endothelial cell (EC) proliferation and growth and the underlying mechanism. Overexpression of HDAC7 by adenoviral gene transfer suppressed human umbilical vein endothelial cell (HUVEC) proliferation by preventing nuclear translocation of beta-catenin and downregulation of T-cell factor-1/Id2 (inhibitor of DNA binding 2) and cyclin D1, leading to G(1) phase elongation. Further assays with the TOPFLASH reporter and quantitative RT-PCR for other beta-catenin target genes such as Axin2 confirmed that overexpression of HDAC7 decreased beta-catenin activity. Knockdown of HDAC7 by lentiviral short hairpin RNA transfer induced beta-catenin nuclear translocation but downregulated cyclin D1, cyclin E1 and E2F2, causing HUVEC hypertrophy. Immunoprecipitation assay and mass spectrometry analysis revealed that HDAC7 directly binds to beta-catenin and forms a complex with 14-3-3 epsilon, zeta, and eta proteins. Vascular endothelial growth factor treatment induced HDAC7 degradation via PLCgamma-IP3K (phospholipase Cgamma-inositol-1,4,5-trisphosphate kinase) signal pathway and partially rescued HDAC7-mediated suppression of proliferation. Moreover, vascular endothelial growth factor stimulation suppressed the binding of HDAC7 with beta-catenin, disrupting the complex and releasing beta-catenin to translocate into the nucleus. These findings demonstrate that HDAC7 interacts with beta-catenin keeping ECs in a low proliferation stage and provides a novel insight into the mechanism of HDAC7-mediated signal pathways leading to endothelial growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
TOPflash (TCF-Reporterplasmid), Transfection grade T-cell factor reporter plasmid containing 2 sets (with the second set in the reverse orientation) of 3 copies of the TCF binding site (wild type) upstream of the Thymidine Kinase minimal promoter and Luciferase open reading frame.
Sigma-Aldrich
IP6K-Inhibitor, The IP6K Inhibitor, also referenced under CAS 519178-28-0, controls the biological activity of IP6K.