Merck
Inhibiting autophagy increases epirubicin's cytotoxicity in breast cancer cells.
Cancer science (2016-08-26)
Wei Guo, Yu Wang, Zhu Wang, Yan-Ping Wang, Hong Zheng
ZUSAMMENFASSUNG
Chemotherapy, radiotherapy, and endocrinotherapy are documented to induce autophagy among breast cancer cells, but the role of autophagy in this disease has been attributed as cytoprotective as well as tumor-suppressing. Thus we studied MDA-MB-231 and SK-BR-3 breast cancer cell lines treated with epirubicin (EPI) to assess autophagy and apoptosis. We found out that EPI induced apoptosis and autophagy in both cell lines. The lysosomal inhibitor bafilomycin A1 inhibited cellular autophagy and enhanced EPI-triggered apoptosis, perhaps due to inhibition of autolysosome formation, which then inhibited autophagic effects of engulfing and clearing damaged mitochondria. This inhibition increased mitochondrial cytochrome C release which augmented epirubicin-induced caspase-dependent apoptosis and cytotoxicity. In addition, the lysosomal neutralizing agent ammonia chloride (AC), and Atg7 knockdown by siRNA, could inhibit epirubicin-triggered autophagy, enhance cytotoxicity, and increase caspase-9- and caspase-3-dependent apoptosis. Thus, autophagy plays a prosurvival role in EPI-treated MDA-MB-231 and SK-BR-3 cells, and autophagy inhibition can potentially reverse this effect and increase the cytotoxicity of EPI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bafilomycin A1 aus Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Anti-Chloride Channel CLC-5 (Clcn5) antibody produced in rabbit, affinity isolated antibody, lyophilized powder