Merck
Experimental diabetic nephropathy is accelerated in matrix metalloproteinase-2 knockout mice.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2012-10-03)
Yoshimi Takamiya, Kei Fukami, Sho-ichi Yamagishi, Yusuke Kaida, Yosuke Nakayama, Nana Obara, Ryuji Iwatani, Ryotaro Ando, Kiyomi Koike, Takanori Matsui, Yuri Nishino, Seiji Ueda, Mark E Cooper, Seiya Okuda
ZUSAMMENFASSUNG
Matrix metalloproteinase-2 (MMP-2) is responsible for the degradation of various types of extracellular matrix (ECM) proteins such as type IV collagen. Decreased MMP-2 expression and activity has been generally thought to contribute to increased accumulation of ECM at the advanced stage of diabetic nephropathy. However, the kinetics and role of MMP-2 in the early phase of diabetic nephropathy remain unclear. To address this issue, we examined whether streptozotocin (STZ)-induced early diabetic nephropathy was accelerated in MMP-2 knockout (KO) mice. Diabetes was induced by the injection of STZ in 6-week-old control and MMP-2 KO mice. Animals were killed after 16 weeks of diabetes of after observation alone. Compared with non-diabetic control mice, renal MMP-2 expression and activity were increased in 16-week old diabetic mice. Serum levels of blood urea nitrogen and creatinine and urinary excretion levels of albumin and N-acetyl-β-D-glucosaminidase were significantly elevated in diabetic MMP-2 KO mice when compared with wild-type diabetic littermates. Further, accumulation of ECM in the glomeruli and atrophy and fibrosis in the tubulointerstitium were exacerbated, and renal α-smooth muscle actin expression was enhanced in diabetic MMP-2 KO mice. Our present study suggests that renal expression and activity of MMP-2 are increased as a compensatory mechanism in the early phase of diabetic nephropathy. Since MMP-2 could play a protective role against the progression of diabetic nephropathy, further enhancement of MMP-2 expression and/or activity in the kidney may be a therapeutic target for the treatment of early diabetic nephropathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid