Merck
  • The chromatin remodeler p400 ATPase facilitates Rad51-mediated repair of DNA double-strand breaks.
The chromatin remodeler p400 ATPase facilitates Rad51-mediated repair of DNA double-strand breaks.
The Journal of cell biology (2012-12-26)
Céline Courilleau, Catherine Chailleux, Alain Jauneau, Fanny Grimal, Sébastien Briois, Elisa Boutet-Robinet, François Boudsocq, Didier Trouche, Yvan Canitrot
ZUSAMMENFASSUNG
DNA damage signaling and repair take place in a chromatin context. Consequently, chromatin-modifying enzymes, including adenosine triphosphate-dependent chromatin remodeling enzymes, play an important role in the management of DNA double-strand breaks (DSBs). Here, we show that the p400 ATPase is required for DNA repair by homologous recombination (HR). Indeed, although p400 is not required for DNA damage signaling, DNA DSB repair is defective in the absence of p400. We demonstrate that p400 is important for HR-dependent processes, such as recruitment of Rad51 to DSB (a key component of HR), homology-directed repair, and survival after DNA damage. Strikingly, p400 and Rad51 are present in the same complex and both favor chromatin remodeling around DSBs. Altogether, our data provide a direct molecular link between Rad51 and a chromatin remodeling enzyme involved in chromatin decompaction around DNA DSBs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture