Merck
  • The MMP-9/TIMP-1 axis controls the status of differentiation and function of myelin-forming Schwann cells in nerve regeneration.
The MMP-9/TIMP-1 axis controls the status of differentiation and function of myelin-forming Schwann cells in nerve regeneration.
PloS one (2012-03-23)
Youngsoon Kim, Albert G Remacle, Andrei V Chernov, Huaqing Liu, Igor Shubayev, Calvin Lai, Jennifer Dolkas, Sergey A Shiryaev, Vladislav S Golubkov, Andrew P Mizisin, Alex Y Strongin, Veronica I Shubayev
ZUSAMMENFASSUNG
Myelinating Schwann cells (mSCs) form myelin in the peripheral nervous system. Because of the works by us and others, matrix metalloproteinase-9 (MMP-9) has recently emerged as an essential component of the Schwann cell signaling network during sciatic nerve regeneration. In the present study, using the genome-wide transcriptional profiling of normal and injured sciatic nerves in mice followed by extensive bioinformatics analyses of the data, we determined that an endogenous, specific MMP-9 inhibitor [tissue inhibitor of metalloproteinases (TIMP)-1] was a top up-regulated gene in the injured nerve. MMP-9 capture followed by gelatin zymography and Western blotting of the isolated samples revealed the presence of the MMP-9/TIMP-1 heterodimers and the activated MMP-9 enzyme in the injured nerve within the first 24 h post-injury. MMP-9 and TIMP-1 co-localized in mSCs. Knockout of the MMP-9 gene in mice resulted in elevated numbers of de-differentiated/immature mSCs in the damaged nerve. Our comparative studies using MMP-9 knockout and wild-type mice documented an aberrantly enhanced proliferative activity and, accordingly, an increased number of post-mitotic Schwann cells, short internodes and additional nodal abnormalities in remyelinated nerves of MMP-9 knockout mice. These data imply that during the first days post-injury MMP-9 exhibits a functionally important anti-mitogenic activity in the wild-type mice. Pharmacological inhibition of MMP activity suppressed the expression of Na(v)1.7/1.8 channels in the crushed nerves. Collectively, our data established an essential role of the MMP-9/TIMP-1 axis in guiding the mSC differentiation and the molecular assembly of myelin domains in the course of the nerve repair process. Our findings of the MMP-dependent regulation of Na(v) channels, which we document here for the first time, provide a basis for therapeutic intervention in sensorimotor pathologies and pain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal, clone 4H8-2, purified from hybridoma cell culture
Sigma-Aldrich
TIMP-1, neutrophile Granulozyten des Menschen, Calbiochem, Native TIMP-1 from human granulocyte.
Sigma-Aldrich
Anti-Sodium Channel PN4 antibody produced in rabbit, affinity isolated antibody, lyophilized powder