Merck
  • Chromosome 19 open reading frame 80 is upregulated by thyroid hormone and modulates autophagy and lipid metabolism.
Chromosome 19 open reading frame 80 is upregulated by thyroid hormone and modulates autophagy and lipid metabolism.
Autophagy (2013-11-23)
Yi-Hsin Tseng, Po-Yuan Ke, Chia-Jung Liao, Sheng-Ming Wu, Hsiang-Cheng Chi, Chung-Ying Tsai, Cheng-Yi Chen, Yang-Hsiang Lin, Kwang-Huei Lin
ZUSAMMENFASSUNG
The thyroid hormone, T 3, regulates cell growth, differentiation and development through binding to the nuclear thyroid hormone receptor (THR), a member of the steroid/TR superfamily of ligand-dependent transcriptional factors. T 3 modulates lipid metabolism in liver, although the detailed molecular mechanisms are unclear at present. Here, by a microarray analysis, we identified a novel chromosome 19 open reading frame 80 (C19orf80) which was activated by T 3. T 3 stimulation led to upregulation of both mRNA and protein levels of C19orf80. Immunofluorescence analysis revealed a vesicle-like pattern of C19orf80 around lipid droplets or within the lysosome-associated compartment in cells. Furthermore, T 3 treatment as well as C19orf80 overexpression specifically activated the autophagic response and lipid metabolism, as observed from lipidated LC3 (LC3-II) and levels of oxygen consumption rate, respectively. Reciprocally, knockdown of C19orf80 obstructed T 3-activated autophagy and lipolysis. Moreover, treatment with autolysosome maturation inhibitors, ammonium chloride and chloroquine, not only suppressed the T 3-activated autophagic process but also lipid metabolism. Our results collectively suggested that T 3 regulates lipid metabolism through a C19orf80-activated autophagic process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
(+)-Etomoxir Natriumsalz Hydrat, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, clone C4, Chemicon®, from mouse