Merck
  • Switch from caspase-dependent to caspase-independent death during heart development: essential role of endonuclease G in ischemia-induced DNA processing of differentiated cardiomyocytes.
Switch from caspase-dependent to caspase-independent death during heart development: essential role of endonuclease G in ischemia-induced DNA processing of differentiated cardiomyocytes.
The Journal of biological chemistry (2006-06-07)
Núria Bahi, Jisheng Zhang, Marta Llovera, Manel Ballester, Joan X Comella, Daniel Sanchis
ZUSAMMENFASSUNG
Differentiated cardiomyocytes are resistant to caspase-dependent cell death; however, the mechanisms involved are still uncertain. We previously reported that low Apaf1 expression partially accounts for cardiomyocyte resistance to apoptosis. Here, we extend the knowledge on the molecular basis of cardiac resistance to caspase activation by showing that the whole caspase-dependent pathway is silenced during heart development. Experimental ischemia triggers caspase activation in embryonic cardiomyocytes and proliferating fibroblasts, but not in neonatal and adult cardiomyocytes. Ischemia induces the release of the proapoptotic factors cytochrome c, truncated-AIF, and EndoG from mitochondria in postnatal cardiomyocytes in the absence of caspase activation. On the one hand, lentiviral-driven knockdown of EndoG shows that this gene is essential for ischemia-induced DNA degradation in neonatal cardiomyocytes, but not in proliferating fibroblasts; on the other hand, the AIF gene is essential for high molecular DNA cleavage in fibroblasts, but not in postmitotic cardiomyocytes, where it plays a prosurvival role during reoxygenation. These results show the switch from caspase-dependent to caspase-independent death pathways after cardiac cell differentiation, and disclose the relevance of EndoG in the caspase-independent DNA processing of differentiated cardiomyocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonales Anti-α-Aktinin (Sarkomer) in Maus hergestellte Antikörper, clone EA-53, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Caspase 12 antibody produced in rat, clone 14F7, purified immunoglobulin, buffered aqueous solution