Merck
  • Let-7 expression is a significant determinant of response to chemotherapy through the regulation of IL-6/STAT3 pathway in esophageal squamous cell carcinoma.
Let-7 expression is a significant determinant of response to chemotherapy through the regulation of IL-6/STAT3 pathway in esophageal squamous cell carcinoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-08-01)
Keijiro Sugimura, Hiroshi Miyata, Koji Tanaka, Rie Hamano, Tsuyoshi Takahashi, Yukinori Kurokawa, Makoto Yamasaki, Kiyokazu Nakajima, Shuji Takiguchi, Masaki Mori, Yuichiro Doki
ZUSAMMENFASSUNG
Cisplatin-based chemotherapy is widely used for esophageal cancer, sometimes in combination with surgery/radiotherapy, but poor response to chemotherapy is not uncommon. The aim of this study was to examine whether miRNA expression is useful to predict the response to chemotherapy in patients with esophageal cancer. Using pretreatment biopsy samples from 98 patients with esophageal cancer who received preoperative chemotherapy, we measured the expression level of several miRNAs whose expression was altered in cisplatin-resistant esophageal cancer cell lines compared with those parent cell lines and examined the relationship between the miRNA expression and response to chemotherapy. In vitro assays were conducted to clarify the mechanism of miRNA-induced changes in chemosensitivity. The expression levels of 15 miRNAs were altered in cisplatin-resistant cells. Of these, low expression of let-7b and let-7c in before-treatment biopsies from 74 patients of the training set correlated significantly with poor response to chemotherapy, both clinically and histopathologically. Low expression of let-7c also correlated with poor prognosis (P = 0.032). The relationship between let-7b and let-7c expression and response to chemotherapy was confirmed in the other 24 patients of the validation set. In in vitro assay, transfection of let-7c restored sensitivity to cisplatin and increased rate of apoptosis after exposure to cisplatin. Let-7c directly repressed cisplatin-activated interleukin (IL)-6/STAT3 prosurvival pathway. Let-7 expression in esophageal cancer can be potentially used to predict the response to cisplatin-based chemotherapy. Let-7 modulates the chemosensitivity to cisplatin through the regulation of IL-6/STAT3 pathway in esophageal cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution