Merck
  • Proteome-wide survey of the autoimmune target repertoire in autoimmune polyendocrine syndrome type 1.
Proteome-wide survey of the autoimmune target repertoire in autoimmune polyendocrine syndrome type 1.
Scientific reports (2016-02-03)
Nils Landegren, Donald Sharon, Eva Freyhult, Åsa Hallgren, Daniel Eriksson, Per-Henrik Edqvist, Sophie Bensing, Jeanette Wahlberg, Lawrence M Nelson, Jan Gustafsson, Eystein S Husebye, Mark S Anderson, Michael Snyder, Olle Kämpe
ZUSAMMENFASSUNG
Autoimmune polyendocrine syndrome type 1 (APS1) is a monogenic disorder that features multiple autoimmune disease manifestations. It is caused by mutations in the Autoimmune regulator (AIRE) gene, which promote thymic display of thousands of peripheral tissue antigens in a process critical for establishing central immune tolerance. We here used proteome arrays to perform a comprehensive study of autoimmune targets in APS1. Interrogation of established autoantigens revealed highly reliable detection of autoantibodies, and by exploring the full panel of more than 9000 proteins we further identified MAGEB2 and PDILT as novel major autoantigens in APS1. Our proteome-wide assessment revealed a marked enrichment for tissue-specific immune targets, mirroring AIRE's selectiveness for this category of genes. Our findings also suggest that only a very limited portion of the proteome becomes targeted by the immune system in APS1, which contrasts the broad defect of thymic presentation associated with AIRE-deficiency and raises novel questions what other factors are needed for break of tolerance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-FAM103A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution