Merck
Substrate ectodomain is critical for substrate preference and inhibition of γ-secretase.
Nature communications (2013-10-11)
Satoru Funamoto, Toru Sasaki, Seiko Ishihara, Mika Nobuhara, Masaki Nakano, Miho Watanabe-Takahashi, Takashi Saito, Nobuto Kakuda, Tomohiro Miyasaka, Kiyotaka Nishikawa, Takaomi C Saido, Yasuo Ihara
ZUSAMMENFASSUNG
Understanding the substrate recognition mechanism of γ-secretase is a key step for establishing substrate-specific inhibition of amyloid β-protein (Aβ) production. However, it is widely believed that γ-secretase is a promiscuous protease and that its substrate-specific inhibition is elusive. Here we show that γ-secretase distinguishes the ectodomain length of substrates and preferentially captures and cleaves substrates containing a short ectodomain. We also show that a subset of peptides containing the CDCYCxxxxCxCxSC motif binds to the amino terminus of C99 and inhibits Aβ production in a substrate-specific manner. Interestingly, these peptides suppress β-secretase-dependent cleavage of APP, but not that of sialyltransferase 1. Most importantly, intraperitoneal administration of peptides into mice results in a significant reduction in cerebral Aβ levels. This report provides direct evidence of the substrate preference of γ-secretase and its mechanism. Our results demonstrate that the ectodomain of C99 is a potent target for substrate-specific anti-Aβ therapeutics to combat Alzheimer's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution