Merck
Transcriptional regulation of Bim by FoxO3A mediates hepatocyte lipoapoptosis.
The Journal of biological chemistry (2007-07-13)
Fernando J Barreyro, Shogo Kobayashi, Steven F Bronk, Nathan W Werneburg, Harmeet Malhi, Gregory J Gores
ZUSAMMENFASSUNG
Hepatocyte lipoapoptosis, a critical feature of nonalcoholic steatohepatitis, can be replicated in vitro by incubating hepatocytes with saturated free fatty acids (FFA). These toxic FFA induce Bim expression, which is requisite for their cytotoxicity. Because the FoxO3a transcription factor has been implicated in Bim expression, our aim was to determine if FFA induce Bim by a FoxO3a-dependent mechanism. In Huh-7 cells, the saturated FFA, palmitic and stearic acid, increased Bim mRNA 16-fold. Treatment of cells with the saturated FFA induced FoxO3a dephosphorylation (activation) and nuclear translocation and stimulated a FoxO luciferase-based reporter assay; direct binding of FoxO3a to the Bim promoter was also confirmed by a chromatin immunoprecipitation assay. A small interfering RNA-targeted knockdown of FoxO3a abrogated FFA-mediated Bim induction and apoptosis. FoxO3a was activated by a phosphatase 2A-dependent mechanism, since okadaic acid- and small interfering RNA-targeted knockdown of this phosphatase blocked FoxO3a dephosphorylation, Bim expression, and apoptosis. Consistent with these data, phosphatase 2A activity was also stimulated 3-fold by saturated FFA. Immunoprecpitation studies revealed an FFA-dependent association between FoxO3a and protein phosphatase 2A. FFA-mediated FoxO3a activation by protein phosphatase 2A was also observed in HepG2 cells and murine hepatocytes. In conclusion, saturated FFA stimulate protein phosphatase 2A activity, which activates FoxO3a, inducing expression of the intracellular death mediator Bim.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Anti-γ-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone GTU-88, ascites fluid
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
JNK-Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
Anti-PP2A-Antikörper, C-Untereinheit, Klon 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Anti-FOXO3a/FKHRL1 Antibody, Upstate®, from rabbit