Merck
  • Activating PIK3CA Mutations Induce an Epidermal Growth Factor Receptor (EGFR)/Extracellular Signal-regulated Kinase (ERK) Paracrine Signaling Axis in Basal-like Breast Cancer.
Activating PIK3CA Mutations Induce an Epidermal Growth Factor Receptor (EGFR)/Extracellular Signal-regulated Kinase (ERK) Paracrine Signaling Axis in Basal-like Breast Cancer.
Molecular & cellular proteomics : MCP (2015-05-09)
Christian D Young, Lisa J Zimmerman, Daisuke Hoshino, Luigi Formisano, Ariella B Hanker, Michael L Gatza, Meghan M Morrison, Preston D Moore, Corbin A Whitwell, Bhuvanesh Dave, Thomas Stricker, Neil E Bhola, Grace O Silva, Premal Patel, Dana M Brantley-Sieders, Maren Levin, Marina Horiates, Norma A Palma, Kai Wang, Philip J Stephens, Charles M Perou, Alissa M Weaver, Joyce A O'Shaughnessy, Jenny C Chang, Ben Ho Park, Daniel C Liebler, Rebecca S Cook, Carlos L Arteaga
ZUSAMMENFASSUNG
Mutations in PIK3CA, the gene encoding the p110α catalytic subunit of phosphoinositide 3-kinase (PI3K) have been shown to transform human mammary epithelial cells (MECs). These mutations are present in all breast cancer subtypes, including basal-like breast cancer (BLBC). Using liquid chromatography-tandem mass spectrometry (LC-MS/MS), we identified 72 protein expression changes in human basal-like MECs with knock-in E545K or H1047R PIK3CA mutations versus isogenic MECs with wild-type PIK3CA. Several of these were secreted proteins, cell surface receptors or ECM interacting molecules and were required for growth of PIK3CA mutant cells as well as adjacent cells with wild-type PIK3CA. The proteins identified by MS were enriched among human BLBC cell lines and pointed to a PI3K-dependent amphiregulin/EGFR/ERK signaling axis that is activated in BLBC. Proteins induced by PIK3CA mutations correlated with EGFR signaling and reduced relapse-free survival in BLBC. Treatment with EGFR inhibitors reduced growth of PIK3CA mutant BLBC cell lines and murine mammary tumors driven by a PIK3CA mutant transgene, all together suggesting that PIK3CA mutations promote tumor growth in part by inducing protein changes that activate EGFR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Anti-Laminin-5-(γ2-Kette-)Antikörper, Klon D4B5, clone D4B5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Eck/EphA2-Antikörper, Klon D7, clone D7, Upstate®, from mouse