Merck
The SIRT1 modulators AROS and DBC1 regulate HSF1 activity and the heat shock response.
PloS one (2013-01-26)
Rachel Raynes, Kathleen M Pombier, Kevin Nguyen, Jessica Brunquell, Jamie E Mendez, Sandy D Westerheide
ZUSAMMENFASSUNG
The heat shock response, the cellular response to protein damaging stress, is critical in maintaining proteostasis. The heat shock response is regulated by the transcription factor HSF1, which is activated upon heat shock and other stresses to induce the expression of molecular chaperones. SIRT1 has previously been shown to activate HSF1 by deacetylating it, leading to increased DNA binding ability. We have investigated how the heat shock response may be controlled by factors influencing SIRT1 activity. We found that heat shock results in an increase in the cellular NAD(+)/NADH ratio and an increase in recruitment of SIRT1 to the hsp70 promoter. Furthermore, we found that the SIRT1 modulators AROS and DBC1 have an impact on hsp70 transcription, HSF1 acetylation status, and HSF1 recruitment to the hsp70 promoter. Therefore, AROS and DBC1 are now two new targets available for therapeutic regulation of the heat shock response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-c-Myc-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Maus-IgG-Antikörper der Ziege, HRP-Konjugat, Upstate®, from goat
Sigma-Aldrich
Anti-Kaninchen-IgG-Antikörper der Ziege, HRP-konjugiert, 1 mg/mL, Upstate®
Sigma-Aldrich
Anti-Sirt1(Sir2)-Antikörper, Upstate®, from rabbit