Merck
Genetic defects in bile acid conjugation cause fat-soluble vitamin deficiency.
Gastroenterology (2013-02-19)
Kenneth D R Setchell, James E Heubi, Sohela Shah, Joel E Lavine, David Suskind, Mohammed Al-Edreesi, Carol Potter, David W Russell, Nancy C O'Connell, Brian Wolfe, Pinky Jha, Wujuan Zhang, Kevin E Bove, Alex S Knisely, Alan F Hofmann, Philip Rosenthal, Laura N Bull
ZUSAMMENFASSUNG
The final step in bile acid synthesis involves conjugation with glycine and taurine, which promotes a high intraluminal micellar concentration to facilitate lipid absorption. We investigated the clinical, biochemical, molecular, and morphologic features of a genetic defect in bile acid conjugation in 10 pediatric patients with fat-soluble vitamin deficiency, some with growth failure or transient neonatal cholestatic hepatitis. We identified the genetic defect that causes this disorder using mass spectrometry analysis of urine, bile, and serum samples and sequence analysis of the genes encoding bile acid-CoA:amino acid N-acyltransferase (BAAT) and bile acid-CoA ligase (SLC27A5). Levels of urinary bile acids were increased (432 ± 248 μmol/L) and predominantly excreted in unconjugated forms (79.4% ± 3.9%) and as sulfates and glucuronides. Glycine or taurine conjugates were absent in the urine, bile, and serum. Unconjugated bile acids accounted for 95.7% ± 5.8% of the bile acids in duodenal bile, with cholic acid accounting for 82.4% ± 5.5% of the total. Duodenal bile acid concentrations were 12.1 ± 5.9 mmol/L, which is too low for efficient lipid absorption. The biochemical profile was consistent with defective bile acid amidation. Molecular analysis of BAAT confirmed 4 different homozygous mutations in 8 patients tested. Based on a study of 10 pediatric patients, genetic defects that disrupt bile acid amidation cause fat-soluble vitamin deficiency and growth failure, indicating the importance of bile acid conjugation in lipid absorption. Some patients developed liver disease with features of a cholangiopathy. These findings indicate that patients with idiopathic neonatal cholestasis or later onset of unexplained fat-soluble vitamin deficiency should be screened for defects in bile acid conjugation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SLC27A5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution