Merck
  • Survival of neural stem cells undergoing DNA damage-induced astrocytic differentiation in self-renewal-promoting conditions in vitro.
Survival of neural stem cells undergoing DNA damage-induced astrocytic differentiation in self-renewal-promoting conditions in vitro.
PloS one (2014-01-30)
Leonid Schneider
ZUSAMMENFASSUNG
We recently reported that neural stem cells (NSCs) become senescent and commit to astrocytic differentiation upon X-ray irradiation. Surprisingly, under self-renewing culture conditions, some of these senescent cells undergo p53-independent apoptosis, which can be suppressed by caspase inhibition and BCL2 overexpression. Inhibition of apoptosis proved beneficial for astroglial differentiation efficiency; hence the toxicity of DNA damage on NSCs was specifically tested in context of the culture conditions. In this regard, self-renewal-promoting culture conditions proved incompatible with terminal astrocyte differentiation and impacted negatively on the viability of NSCs following DNA damage-induced cell cycle exit. On the contrary, a switch to differentiation-supporting conditions ablated apoptosis and conveyed tolerance to DNA damage. Thus, stem cell death has likely not originated from DNA break toxicity, while the potentially confounding effect of stem cell niche should always be taken in consideration in stem cell irradiation experiments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid