Merck
Wnt4 signaling prevents skeletal aging and inflammation by inhibiting nuclear factor-κB.
Nature medicine (2014-08-12)
Bo Yu, Jia Chang, Yunsong Liu, Jiong Li, Kareena Kevork, Khalid Al-Hezaimi, Dana T Graves, No-Hee Park, Cun-Yu Wang
ZUSAMMENFASSUNG
Aging-related bone loss and osteoporosis affect millions of people worldwide. Chronic inflammation associated with aging promotes bone resorption and impairs bone formation. Here we show that Wnt4 attenuates bone loss in osteoporosis and skeletal aging mouse models by inhibiting nuclear factor-κB (NF-κB) via noncanonical Wnt signaling. Transgenic mice expressing Wnt4 from osteoblasts were significantly protected from bone loss and chronic inflammation induced by ovariectomy, tumor necrosis factor or natural aging. In addition to promoting bone formation, Wnt4 inhibited osteoclast formation and bone resorption. Mechanistically, Wnt4 inhibited NF-κB activation mediated by transforming growth factor-β-activated kinase-1 (Tak1) in macrophages and osteoclast precursors independently of β-catenin. Moreover, recombinant Wnt4 alleviated bone loss and inflammation by inhibiting NF-κB in vivo in mouse models of bone disease. Given its dual role in promoting bone formation and inhibiting bone resorption, our results suggest that Wnt4 signaling could be an attractive therapeutic target for treating osteoporosis and preventing skeletal aging.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-NFκB p65 Antibody, CT, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-TBP in Maus hergestellte Antikörper, clone 58C9, purified from hybridoma cell culture