Merck
  • Tasquinimod Is an Allosteric Modulator of HDAC4 survival signaling within the compromised cancer microenvironment.
Tasquinimod Is an Allosteric Modulator of HDAC4 survival signaling within the compromised cancer microenvironment.
Cancer research (2012-11-15)
John T Isaacs, Lizamma Antony, Susan L Dalrymple, W Nathaniel Brennen, Stephanie Gerber, Hans Hammers, Michel Wissing, Sushant Kachhap, Jun Luo, Li Xing, Per Björk, Anders Olsson, Anders Björk, Tomas Leanderson
ZUSAMMENFASSUNG
Tasquinimod is an orally active antiangiogenic drug that is currently in phase III clinical trials for the treatment of castration-resistant prostate cancer. However, the target of this drug has remained unclear. In this study, we applied diverse strategies to identify the histone deacetylase HDAC4 as a target for the antiangiogenic activity of tasquinimod. Our comprehensive analysis revealed allosteric binding (Kd 10-30 nmol/L) to the regulatory Zn(2+) binding domain of HDAC4 that locks the protein in a conformation preventing HDAC4/N-CoR/HDAC3 complex formation. This binding inhibited colocalization of N-CoR/HDAC3, thereby inhibiting deacetylation of histones and HDAC4 client transcription factors, such as HIF-1α, which are bound at promoter/enhancers where epigenetic reprogramming is required for cancer cell survival and angiogenic response. Through this mechanism, tasquinimod is effective as a monotherapeutic agent against human prostate, breast, bladder, and colon tumor xenografts, where its efficacy could be further enhanced in combination with a targeted thapsigargin prodrug (G202) that selectively kills tumor endothelial cells. Together, our findings define a mechanism of action of tasquinimod and offer a perspective on how its clinical activity might be leveraged in combination with other drugs that target the tumor microenvironment. Cancer Res; 73(4); 1386-99. ©2012 AACR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Acetyl-Histon-H3-Antikörper, from rabbit
Millipore
Affinitätsgel ANTI-FLAG® M1 Agarose
Sigma-Aldrich
Anti-Acetyl-Lysin-Antikörper, serum, Upstate®