Merck
Akt contributes to neuroprotection by hypothermia against cerebral ischemia in rats.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2005-10-21)
Heng Zhao, Takayoshi Shimohata, Jade Q Wang, Guohua Sun, David W Schaal, Robert M Sapolsky, Gary K Steinberg
ZUSAMMENFASSUNG
Activation of the Akt/protein kinase B (PKB) kinase pathway can be neuroprotective after stroke. Akt is activated by growth factors via a phosphorylation-dependent pathway involving the kinases phosphoinositide 3 (PI3) kinase and phosphoinositide-dependent protein kinase-1 (PDK1) and is negatively regulated by phosphatase and tensin homolog deleted on chromosome 10 (PTEN). Akt kinase blocks apoptosis by phosphorylating the substrates forkhead transcription factor (FKHR) and glycogen synthase kinase 3beta (GSK3beta). We found that intra-ischemic hypothermia (30 degrees C) reduced infarct size and improved functional outcomes up to 2 months. Changes in phosphorylation levels of Akt, as measured by Western blots and immunostaining, differed from levels of Akt activity measured in an in vitro assay in normothermic animals. Hypothermia blocked most of these changes and maintained Akt activity. Inhibition of PI3/Akt enlarged infarct size in hypothermic animals. Hypothermia improved phosphorylation of PDK1, PTEN, and FKHR. Hypothermia did not improve GSK3beta (Ser9) phosphorylation but blocked the nuclear translocation of phosphorylated beta-catenin (Ser33/37/Thr41) downstream of GSK3beta. Phosphorylation levels of PTEN, Akt, and Akt substrate decreased before apoptotic cytochrome c release and degradation of microtubule-associated protein-2, a marker of neuronal survival. Hypothermia may protect from ischemic damage in part by preserving Akt activity and attenuating the apoptotic effects of PTEN, PDK1, and FKHR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-MAP2 in Maus hergestellte Antikörper, ascites fluid, clone HM-2