Merck
  • Structure-function studies of Escherichia coli RpoH (sigma32) by in vitro linker insertion mutagenesis.
Structure-function studies of Escherichia coli RpoH (sigma32) by in vitro linker insertion mutagenesis.
Journal of bacteriology (2003-04-18)
Franz Narberhaus, Sylvia Balsiger
ZUSAMMENFASSUNG
The sigma factor RpoH (sigma(32)) is the key regulator of the heat shock response in Escherichia coli. Many structural and functional properties of the sigma factor are poorly understood. To gain further insight into RpoH regions that are either important or dispensable for its cellular activity, we generated a collection of tetrapeptide insertion variants by a recently established in vitro linker insertion mutagenesis technique. Thirty-one distinct insertions were obtained, and their sigma factor activity was analyzed by using a groE-lacZ reporter fusion in an rpoH-negative background. Our study provides a map of permissive sites which tolerate linker insertions and of functionally important regions at which a linker insertion impairs sigma factor activity. Selected linker insertion mutants will be discussed in the light of known sigma factor properties and in relation to a modeled structure of an RpoH fragment containing region 2.