- Oncogenic function of p34SEI-1 via NEDD4‑1‑mediated PTEN ubiquitination/degradation and activation of the PI3K/AKT pathway.
Oncogenic function of p34SEI-1 via NEDD4‑1‑mediated PTEN ubiquitination/degradation and activation of the PI3K/AKT pathway.
International journal of oncology (2013-08-24)
Samil Jung, Chengping Li, Dongjun Jeong, Soonduck Lee, Jiyeon Ohk, Meeyoung Park, Songyi Han, Jingjing Duan, Changjin Kim, Young Yang, Keun-Il Kim, Jong-Seok Lim, Young-Sook Kang, Myeong-Sok Lee
PMID23970032
ZUSAMMENFASSUNG
A 34-KD protein encoded by the SEI-1 gene (p34(SEI‑1)), is a relatively recently discovered oncoprotein that has multiple important biological functions. Our data show that p34(SEI-1) enhances cancer cell survival and promotes tumorigenesis by downregulating the tumor suppressor PTEN, a negative regulator of the PI3K/AKT signaling pathway, and therefore activating the PI3K/AKT signaling pathway. In this process, p34(SEI-1) positively affects NEDD4-1 gene expression both at the transcriptional and protein levels. Furthermore, the expression levels of p34(SEI-1) and NEDD4-1 were found to be coordinated in tumor tissues obtained from patients with breast cancer. We also show that p34(SEI-1) affects the subcellular localization of PTEN.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid