Merck
  • Human vascular endothelial growth factor protects axotomized retinal ganglion cells in vivo by activating ERK-1/2 and Akt pathways.
Human vascular endothelial growth factor protects axotomized retinal ganglion cells in vivo by activating ERK-1/2 and Akt pathways.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2006-12-01)
Ulkan Kilic, Ertugrul Kilic, Anne Järve, Zeyun Guo, Annett Spudich, Katja Bieber, Uxue Barzena, Claudio L Bassetti, Hugo H Marti, Dirk M Hermann
ZUSAMMENFASSUNG
Based on its trophic effects on neurons and vascular cells, vascular endothelial growth factor (VEGF) is a promising candidate for the treatment of neurodegenerative diseases. To evaluate the therapeutic potential of VEGF, we here examined effects of this growth factor on the degeneration of axotomized retinal ganglion cells (RGCs), which, as CNS-derived neurons, offer themselves in an excellent way to study neuroprotection in vivo. Making use of a transgenic mouse line that constitutively expresses human VEGF under a neuron-specific enolase promoter, we show that (1) the VEGF-transgenic retina overexpresses human VEGF, (2) RGCs carry the VEGF receptor-2, and (3) vascular networks in normal and axotomized VEGF-transgenic (tg) retinas do not differ from control animals. After axotomy, RGCs of VEGF-tg mice were protected against delayed degeneration, as compared with wild-type littermates. Western blots revealed increased phosphorylated ERK-1/2 and Akt and reduced phosphorylated p38 and activated caspase-3 levels in axotomized VEGF-transgenic retinas. Intravitreous injections of pharmacological ERK-1/2 (PD98059) or Akt (LY294002) inhibitors showed that VEGF exerts neuroprotection by dual activation of ERK-1/2 and Akt pathways. In view that axotomy-induced RGC death occurs slowly and considering that RGCs are CNS-derived neurons, we predict the clinical implementation of VEGF in neurodegenerative diseases of both brain and retina.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
Monoklonale Anti-MAP-Kinase, aktiviert (Diphosphorylierter ERK-1&2) in Maus hergestellte Antikörper, clone MAPK-YT, ascites fluid