Merck
  • Increased expression of oxidation-specific epitopes and apoptosis are associated with haptoglobin genotype: possible implications for plaque progression in human atherosclerosis.
Increased expression of oxidation-specific epitopes and apoptosis are associated with haptoglobin genotype: possible implications for plaque progression in human atherosclerosis.
Journal of the American College of Cardiology (2012-07-07)
K-Raman Purushothaman, Meerarani Purushothaman, Andrew P Levy, Patrick A Lento, Solene Evrard, Jason C Kovacic, Karen C Briley-Saebo, Sotirios Tsimikas, Joseph L Witztum, Prakash Krishnan, Annapoorna Kini, Zahi A Fayad, Valentin Fuster, Samin K Sharma, Pedro R Moreno
ZUSAMMENFASSUNG
The purpose of this study was to test the hypothesis that increased oxidative stress is associated with apoptosis in human plaques with the haptoglobin (Hp) 2-2 genotype. Intraplaque hemorrhage releases free hemoglobin (Hb). Impaired Hb clearance induces oxidative stress leading to plaque progression. The binding of Hp to Hb attenuates iron-induced oxidative reactions. Twenty-six human aortic plaques were Hp genotyped. Hp2-2 plaques (n = 13) were compared with control (Hp1-1/2-1) (n = 13). The iron grade was measured by Perl's staining. Immunostaining was used to detect oxidation-specific epitopes (OSEs) reflecting oxidized phospholipids and malondialdehyde-like epitopes. The percentages of apoptotic cells and apoptotic morphological features were quantified. DNA fragmentation and active caspase-3 were measured by in situ end-labeling and immunohistochemistry, respectively. In Hp2-2 plaques, iron content was increased (1.22 ± 0.15 vs. 0.54 ± 0.08; p < 0.0001) along with expression of oxidized phospholipid- (78.9 ± 5.8 vs. 38.8 ± 3.8; p < 0.0001), and malondialdehyde-like OSEs (93.9 ± 7.9 vs. 54.7 ± 3.9; p < 0.0001). The total percentages of apoptotic cells (11.9 ± 0.44 vs. 3.5 ± 0.28; p < 0.0001), nuclear fragmentation (11.8 ± 0.50 vs. 3.3 ± 0.26; p < 0.0001), nuclear condensation (10.9 ± 0.58 vs. 3.4 ± 0.20; p < 0.0001), chromatin margination (14.2 ± 0.57 vs. 6.5 ± 0.37; p < 0.0001), cytoplasmic blebs (1.6 ± 0.28 vs. 0.8 ± 0.14; p < 0.002), and eosinophilia (10.8 ± 0.74 vs. 4.2 ± 0.27; p < 0.0001) were increased in Hp2-2 plaques. Furthermore, DNA fragmentation (119.9 ± 1.40 vs. 57.5 ± 0.80; p < 0.001), and active caspase-3 density (84.7 ± 7.62 vs. 50.6 ± 7.49; p < 0.004) were increased in Hp2-2 plaques. Logistic regression analysis identified correlation between the percentage of apoptotic cells and the density of OSEs (r = 0.56; p < 0.003). These findings provide insights into genetic predisposition to oxidative stress and the relationship between OSEs and macrophage apoptosis that may explain advanced atherosclerosis in human Hp2-2 plaques.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin α-Glattmuskel-FITC-Antikörper der Maus in Maus hergestellte Antikörper, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human Kappa Light Chains (Bound and Free)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution