Merck
Cell polarity defines three distinct domains in pancreatic β-cells.
Journal of cell science (2016-02-28)
Wan J Gan, Michael Zavortink, Christine Ludick, Rachel Templin, Robyn Webb, Richard Webb, Wei Ma, Philip Poronnik, Robert G Parton, Herbert Y Gaisano, Annette M Shewan, Peter Thorn
ZUSAMMENFASSUNG
The structural organisation of pancreatic β-cells in the islets of Langerhans is relatively unknown. Here, using three-dimensional (3D) two-photon, 3D confocal and 3D block-face serial electron microscopy, we demonstrate a consistent in situ polarisation of β-cells and define three distinct cell surface domains. An apical domain located at the vascular apogee of β-cells, defined by the location of PAR-3 (also known as PARD3) and ZO-1 (also known as TJP1), delineates an extracellular space into which adjacent β-cells project their primary cilia. A separate lateral domain, is enriched in scribble and Dlg, and colocalises with E-cadherin and GLUT2 (also known as SLC2A2). Finally, a distinct basal domain, where the β-cells contact the islet vasculature, is enriched in synaptic scaffold proteins such as liprin. This 3D analysis of β-cells within intact islets, and the definition of distinct domains, provides new insights into understanding β-cell structure and function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Insulin-Antikörper in Maus hergestellte Antikörper, clone K36AC10, ascites fluid
Sigma-Aldrich
Anti-PARD3-Antikörper, Upstate®, from rabbit