- Short report: detection of 72-75-kD and 123-kD fractions of Leishmania antigen in urine of patients with visceral leishmaniasis.
Short report: detection of 72-75-kD and 123-kD fractions of Leishmania antigen in urine of patients with visceral leishmaniasis.
The American journal of tropical medicine and hygiene (1995-05-01)
M De Colmenares, M Portus, C Riera, M Gallego, M J Aisa, S Torras, C Munoz
PMID7539596
ZUSAMMENFASSUNG
Two polypeptide fractions of 72-75 kD were detected in the urine of 14 of 15 patients with visceral leishmaniasis (VL) and another fraction of 123 kD was found in 10 of the 15 patients by using a Western blot technique. None of these fractions was detected in the urine of 20 controls. These results suggest that antigen detection in urine could be a powerful, noninvasive method for VL diagnosis.