Merck
  • Downregulated endogenous sulfur dioxide/aspartate aminotransferase pathway is involved in angiotensin II-stimulated cardiomyocyte autophagy and myocardial hypertrophy in mice.
Downregulated endogenous sulfur dioxide/aspartate aminotransferase pathway is involved in angiotensin II-stimulated cardiomyocyte autophagy and myocardial hypertrophy in mice.
International journal of cardiology (2016-10-23)
Qinghua Chen, Lulu Zhang, Siyao Chen, Yaqian Huang, Kun Li, Xiaoqi Yu, Huijuan Wu, Xiaoyu Tian, Chunyu Zhang, Chaoshu Tang, Junbao Du, Hongfang Jin
ZUSAMMENFASSUNG
The study was designed to investigate if endogenous sulfur dioxide (SO2) was involved in cardiomyocyte autophagy and myocardial hypertrophy stimulated by angiotensin II (Ang II). Thirty-two C57 mice were randomly divided into control, SO2, Ang II and Ang II+SO2 groups. Human myocardial cell line H9c2 was divided into four groups including control, SO2, Ang II and Ang II+SO2 groups. Blood pressure and myocardial hypertrophy of the mice were measured two weeks after Ang II administration. LC3 II/I ratio, and Beclin1, Atg4B and p62 expressions were determined both in vivo and in vitro. Autophagosome was observed in H9c2 cells with confocal microscope. Endogenous SO2 generation and aspartate aminotransferase (AAT) expression were measured. In animal studies, hypertension and myocardial hypertrophy developed two weeks after Ang II administration. LC3 II/I ratio and Beclin1 and Atg4B expressions were markedly elevated (P all <0.05), but p62 expression was lowered (P<0.05) both in vivo and in vitro. Compared with control group, endogenous SO2 levels, AAT activity and AAT2 expression were obviously down-regulated (P all <0.05). However, SO2 donor significantly reduced Ang II-induced myocardial hypertrophy in mice. LC3 II/I ratio and Beclin1 and Atg4B expressions were down-regulated (P all <0.05) but p62 expression was significantly increased (P<0.05) in the presence of SO2 both in vivo and in vitro. Down-regulated endogenous SO2/AAT2 pathway might be involved in the pathogenesis of myocardial hypertrophy. SO2 prevented Ang II -induced myocardial hypertrophy accompanied by down-regulating cardiomyocyte autophagy.