Merck
  • Icariin promotes osteogenic differentiation of rat bone marrow stromal cells by activating the ERα-Wnt/β-catenin signaling pathway.
Icariin promotes osteogenic differentiation of rat bone marrow stromal cells by activating the ERα-Wnt/β-catenin signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-10-21)
QiuShi Wei, Jin Zhang, GuoJu Hong, ZhenQiu Chen, WeiMin Deng, Wei He, Mei Hui Chen
ZUSAMMENFASSUNG
Icariin, the main effective component of Herba Epimedii, has been identified to regulate the osteogenic differentiation of rat bone marrow stromal cells (rBMSCs) and have potential application in preventing bone loss and promoting bone regeneration. However, the underlying signaling pathway of its osteogenic effect remains unclear. Here, we investigated whether icariin could promote osteogenesis of rBMSCs through modulating the estrogen receptor alpha (ERα) and Wnt/β-catenin signaling pathways, which implicated in rBMSCs osteogenesis. rBMSCs were cultured in osteogenic induction medium and treated with icariin or together with ICI 182780 or DKK1, the effects on the expression of osteogenic genes and Wnt/β-catenin signaling were detected. Results indicated that icariin (0.1μM) markedly enhanced the proliferation of rBMSCs and alkaline phosphatase (ALP) activity. Additionally, icariin (0.1μM) significantly up-regulated the expression of osteogenic genes (Runx2, osteopotin, DLX5, osteocalcin, collagen type I, and ERα) and Wnt signal members (β-catenin, Lef1, TCF7, c-jun, c-myc, and cyclin D). Furthermore, icariin stimulated the activation of β-catenin as evidenced by increased total β-catenin protein and nuclear translocation. These osteogenesis-potentiating effects of icariin were blocked by ICI 182780 or DKK1. Taken together, these results suggest that the osteogenic effects of icariin on rBMSCs involves the ERα-Wnt/β-catenin signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)