Merck
  • Autophagy induced by AXL receptor tyrosine kinase alleviates acute liver injury via inhibition of NLRP3 inflammasome activation in mice.
Autophagy induced by AXL receptor tyrosine kinase alleviates acute liver injury via inhibition of NLRP3 inflammasome activation in mice.
Autophagy (2016-10-27)
Jihye Han, Joonbeom Bae, Chang-Yong Choi, Sang-Pil Choi, Hyung-Sik Kang, Eun-Kyeong Jo, Jongsun Park, Young Sik Lee, Hyun-Seuk Moon, Chung-Gyu Park, Myung-Shik Lee, Taehoon Chun
ZUSAMMENFASSUNG
Severe hepatic inflammation is a common cause of acute or chronic liver disease. Macrophages are one of the key mediators which regulate the progress of hepatic inflammation. Increasing evidence shows that the TAM (TYRO3, AXL and MERTK) family of RTKs (receptor tyrosine kinases), which is expressed in macrophages, alleviates inflammatory responses through a negative feedback loop. However, the functional contribution of each TAM family member to the progression of hepatic inflammation remains elusive. In this study, we explore the role of individual TAM family proteins during autophagy induction and evaluate their contribution to hepatic inflammation. Among the TAM family of RTKs, AXL (AXL receptor tyrosine kinase) only induces autophagy in macrophages after interaction with its ligand, GAS6 (growth arrest specific 6). Based on our results, autophosphorylation of 2 tyrosine residues (Tyr815 and Tyr860) in the cytoplasmic domain of AXL in mice is required for autophagy induction and AXL-mediated autophagy induction is dependent on MAPK (mitogen-activated protein kinase)14 activity. Furthermore, induction of AXL-mediated autophagy prevents CASP1 (caspase 1)-dependent IL1B (interleukin 1, β) and IL18 (interleukin 18) maturation by inhibiting NLRP3 (NLR family, pyrin domain containing 3) inflammasome activation. In agreement with these observations, axl-/- mice show more severe symptoms than do wild-type (Axl+/+) mice following acute hepatic injury induced by administration of lipopolysaccharide (LPS) or carbon tetrachloride (CCl4). Hence, GAS6-AXL signaling-mediated autophagy induction in murine macrophages ameliorates hepatic inflammatory responses by inhibiting NLRP3 inflammasome activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatasehemmer-Cocktail 3, DMSO solution
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Neomycin -trisulfat (Salz) Hydrat, powder
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid
Sigma-Aldrich
Chloroquin -diphosphat (Salz), powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Hexadimethrinbromid, ≥94% (titration)
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Paraformaldehyd, reagent grade, crystalline
Sigma-Aldrich
Formalinlösung, neutral gepuffert, 10 %, case of 48 × 15 mL, histological tissue fixative
Sigma-Aldrich
Geys ausgewogene Salzlösung, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Glybenclamid, ≥99% (HPLC)
Millipore
Protease-Inhibitor-Cocktail-Set III, frei von tierischen Bestandteilen, Protease Inhibitor Cocktail Set III, Animal-Free, is a cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine & serine proteases & aminopeptidases.