- A Near Infrared Fluorescent Probe for Sensitive Determination of Human Serum Albumin.
A Near Infrared Fluorescent Probe for Sensitive Determination of Human Serum Albumin.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2016-12-13)
Xiaodan Zeng, Mingshuo Ma, Baocun Zhu, Lin Zhu
PMID27941257
ZUSAMMENFASSUNG
A fluorescent probe 1 has been successfully developed to determine human serum albumin (HSA). Probe 1 expresses a dramatic fluorescence enhancement to HSA without interference from other amino acids. Under the optimal conditions, the calibration graphs are linear over the range of 0 - 13.3 μg/mL with the limit of determination of 0.61 μg/mL. Thus, this probe shows high sensitivity and selectivity to HSA.