Merck
Essential oil from fruit of Xylopia langsdorffiana: antitumour activity and toxicity.
Pharmaceutical biology (2016-08-26)
Ana Paula Gomes Moura, Daiene Martins Beltrão, João Carlos Lima Rodrigues Pita, Aline Lira Xavier, Monalisa Taveira Brito, Tatyanna Kelvia Gomes de Sousa, Leônia Maria Batista, João Ernesto de Carvalho, Ana Lúcia Tasca Gois Ruiz, Adriana Della Torre, Marcelo Cavalcante Duarte, Josean Fechine Tavares, Marcelo Sobral da Silva, Marianna Vieira Sobral
ZUSAMMENFASSUNG
The genus Xylopia L. (Annonaceae) includes aromatic plants that have both nutritional and medicinal uses. Essential oils of Xylopia species have antitumour effects. However, the efficacy of the essential oil from the fruit of Xylopia langsdorffiana St. Hil & Tul. (EOX) has not been examined. EOX was evaluated to determine its chemical composition, antitumour activity and toxicity. EOX was obtained from fresh fruits of X. langsdorffiana subjected to hydrodistillation, and gas chromatography-mass spectrometry was used to characterize the chemical composition of EOX. The toxicity of EOX was evaluated using haemolysis, acute toxicity and micronucleus assays. The in vitro antitumour activity of EOX was investigated using the sulforhodamine B assay. The sarcoma 180 murine tumour model was used to evaluate the in vivo antitumour activity and toxicity of EOX (50 and 100 mg/kg) after 7 d of treatment. The major components of EOX were α-pinene (34.57%) and limonene (31.75%). The HC50 (concentration producing 50% haemolysis) was 293.6 μg/ml. EOX showed greater selectivity for the leukaemia cell line K562, with total growth inhibition (TGI) (concentration producing TGI) of 1.8 μg/ml, and for multidrug-resistant ovarian tumour cell line NCI/ADR-RES (TGI of 45.4 μg/ml). The LD50 was approximately 351.09 mg/kg. At doses of 50 and 100 mg/kg, EOX inhibited the in vivo growth of sarcoma 180 by 38.67 and 54.32%, respectively. EOX displayed minor hepatic alterations characteristic of acute hepatitis and induced no genotoxicity. EOX showed in vitro and in vivo antitumour activity and low toxicity, which warrants further pharmacological studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade