Merck
  • Reduced apurinic/apyrimidinic endonuclease activity enhances the antitumor activity of oxymatrine in lung cancer cells.
Reduced apurinic/apyrimidinic endonuclease activity enhances the antitumor activity of oxymatrine in lung cancer cells.
International journal of oncology (2016-10-18)
Zhiqiang Wang, Wenya Xu, Ziying Lin, Chunyan Li, Yahong Wang, Lawei Yang, Gang Liu
ZUSAMMENFASSUNG
Lung cancer is the leading cause of cancer-related deaths worldwide and is associated with a very poor outcome. Oxymatrine exerts antitumor effects by inducing apoptosis and inhibiting the proliferation of different cancer cells; however, the anticancer effects and mechanism of action of oxymatrine have not been evaluated sufficiently in human lung cancer cells. Thus, the present study aimed to investigate the anticancer effects of oxymatrine in human lung cancer cells and identify the molecular mechanisms underlying these effects. MTT assays demonstrated that oxymatrine significantly inhibited the proliferation of A549 and H1299 cells in a time- and dose-dependent manner. In addition, flow cytometry and terminal deoxynucleotidyl transferase-mediated dUTP-biotin nick end-labeling assays suggested that oxymatrine treatment may induce lung cancer cell apoptosis in a dose-dependent manner. Furthermore, we detected that oxymatrine induced a significant increase in DNA damage and the expression of PARP and phosphorylated H2AX, and a significant decrease in that of nuclear APE1 and AP endonuclease activity in A549 cells. APE1 knockdown cells (APE1shRNA) plus oxymatrine treatment reduced cells proliferation and induced apoptosis more seriously than control shRNA cells. This appeared to be a consequence of an increase in the number of apurinic/apyrimidinic (AP) sites, DNA damage, PARP and H2AX phosphorylation, which together resulted in the induction of apoptosis. In contrast, the sensitizing effects of APE1 overexpression plus oxymatrine treatment did not occur in APEOE cells. These findings reveal a potential mechanism of action for oxymatrine-induced apoptosis and suggest that oxymatrine is a promising potential therapeutic agent for the treatment of lung cancer.