Merck
  • Total and single species of uronic acid-bearing glycosaminoglycans in urine of newborns of 2-3days of age for early diagnosis application.
Total and single species of uronic acid-bearing glycosaminoglycans in urine of newborns of 2-3days of age for early diagnosis application.
Clinica chimica acta; international journal of clinical chemistry (2016-11-05)
Francesca Maccari, Fabio Galeotti, Lucia Zampini, Lucia Padella, Rosella Tomanin, Daniela Concolino, Agata Fiumara, Tiziana Galeazzi, Giovanni Coppa, Orazio Gabrielli, Nicola Volpi
ZUSAMMENFASSUNG
Urine are easily accessible and relatively simple to process and uronic acid-bearing glycosaminoglycans (UA-GAGs) may serve as biomarkers for several diseases, like for mucopolysaccharidosis. We report a study from a large cohort of healthy newborns of 2-3days to have a basic profile of total content of urinary UA-GAGs, their composition and structural signatures utilizing a rapid extractive method and sensitive separation of enzymatic released disaccharides by capillary electrophoresis-light induced fluorescence. Results were also compared with those obtained from normal adult subjects. A total of UA-GAGs content of ~35μg/mg creatinine was observed in 331 newborns versus 1.5μg/mg creatinine of adult urine composed of ~90% chondroitin sulfate (CS), ~7% heparan sulfate (HS) and ~3% hyaluronic acid (HA). No significant differences were observed with adults. Specific ratios between the main CS disaccharides were informative of a significant greater 4-sulfation and charge density for newborn compared to adults. The HS from newborn urine was mainly composed by the non-sulfated (~64%) and mono-sulfated (~28%) disaccharides. No significant differences were observed versus adult urine. The present method is able to measure changes in UA-GAG composition and their structure independently of the age of subjects and rapidly applicable to the newborn diagnosis without necessity to have creatinine levels. Moreover, modifications in charge density values as well as the presence of sulfate groups in specific positions may be indicative of altered conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminoacridon, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)