Merck
  • Effect of early administration of lower dose versus high dose of fresh mitochondria on reducing monocrotaline-induced pulmonary artery hypertension in rat.
Effect of early administration of lower dose versus high dose of fresh mitochondria on reducing monocrotaline-induced pulmonary artery hypertension in rat.
American journal of translational research (2017-01-13)
Tien-Hung Huang, Sheng-Ying Chung, Sarah Chua, Han-Tan Chai, Jiunn-Jye Sheu, Yi-Ling Chen, Chih-Hung Chen, Hsueh-Wen Chang, Meng-Shen Tong, Pei-Hsun Sung, Cheuk-Kwan Sun, Hung-I Lu, Hon-Kan Yip
ZUSAMMENFASSUNG
This study aim to investigate whether early mitochondrial administration would be effective and whether high-dose mitochondria (15000 μg/rat) would be more effective than low-dose mitochondria (1500 μg/rat) for attenuating the monocrotaline (MCT/65 mg/kg/rat)-induced pulmonary artery hypertension (PAH) in rat. Male-adult SD rats (n = 32) were randomized categorized into groups 1 (sham-control), 2 (PAH), 3 (PAH + low-dose mitochondria), and 4 (PAH + high-dose mitochondria). Mitochondria were admitted at day 5 and rats were sacrificed at day 35 post-MCT treatment. By day 35, oxygen saturation (saO2) was highest in group 1 and lowest in group 2, and significantly higher in group 3 than in group 4 (P<0.001). Conversely, right ventricular systolic blood pressure showed an opposite pattern compared with saO2 among all groups (P<0.001). Histological integrity of alveolar sacs exhibited a pattern identical to saO2, whereas lung crowding score and number of muscularized artery displayed an opposite pattern (all P<0.001). The protein expression of indices of inflammation (MMP-9, TNF-α, NF-κB), oxidative stress (oxidized protein, NO-1, NOX-2, NOX-4), apoptosis (Bax, cleaved caspase-3 and PARP), fibrosis (p-Smad3, TGF-β), mitochondrial-damage (cytosolic cytochrome-C), and hypoxia-smooth muscle proliferative factors (HIF-α, connexin43, TRPCs) showed an opposite pattern compared, whereas anti-fibrosis (p-Smad1/5, BMP-2) and mitochondrial integrity (mitochondrial cytochrome-C) exhibited an identical pattern to saO2 in all groups (all P<0.001). Low dose is superior to high dose of mitochondria for protecting against MCT-induced PAH. The paradoxical beneficial effect may imply therapy with 15000 μg/rat mitochondria is overdose in this situation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
OxyBlot Proteinoxidations-Nachweiskit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Adenosin-5′-diphosphoribose Natriumsalz, ≥93%