Merck
  • Dehydrative direct arylations of arenes with phenols via ruthenium-catalyzed C-H and C-OH bond functionalizations.
Dehydrative direct arylations of arenes with phenols via ruthenium-catalyzed C-H and C-OH bond functionalizations.
Organic letters (2008-10-16)
Lutz Ackermann, Michael Mulzer
ZUSAMMENFASSUNG
Phenols can be employed as proelectrophiles in operationally simple ruthenium-catalyzed dehydrative direct arylations, proceeding through chemo- and regioselective functionalizations of C-H and C-OH bonds.