Merck
  • Effects of dentifrices differing in fluoride compounds on artificial enamel caries lesions in vitro.
Effects of dentifrices differing in fluoride compounds on artificial enamel caries lesions in vitro.
Odontology (2016-02-06)
R J Wierichs, H Zelck, C E Doerfer, P Appel, S Paris, M Esteves-Oliveira, H Meyer-Lueckel
ZUSAMMENFASSUNG
The aim of this study was to compare the caries-preventive effect of a stabilized stannous fluoride/sodium fluoride dentifrice containing sodium hexametaphosphate with those of a regular, solely sodium fluoride-containing and amine fluoride-containing dentifrice on pre-demineralized bovine enamel specimens using a pH-cycling model. Bovine enamel specimens with two artificial lesions each were prepared. Baseline mineral loss of both lesions was analyzed using transversal microradiography (TMR). Eighty-five specimens with a mean (SD) baseline mineral loss of 3393 (683) vol% × µm were selected and randomly allocated to five groups (n = 13/15). Treatments during pH-cycling (28 days and 2 × 20 min demineralization/day) were: brushing twice daily with slurries of AmF (1400 ppm F
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D.E.R. 332, used as embedding medium