Merck
  • Cytotoxic effects of nanosilver are highly dependent on the chloride concentration and the presence of organic compounds in the cell culture media.
Cytotoxic effects of nanosilver are highly dependent on the chloride concentration and the presence of organic compounds in the cell culture media.
Journal of nanobiotechnology (2017-01-08)
Jean-Pierre Kaiser, Matthias Roesslein, Liliane Diener, Adrian Wichser, Bernd Nowack, Peter Wick
ZUSAMMENFASSUNG
Nanosilver shows great promise for use in industrial, consumer or medical products because of its antimicrobial properties. However, the underlying mechanisms of the effects of silver nanoparticles on human cells are still controversial. Therefore, in the present study the influence of the chloride concentration and different serum content of culture media on the cytotoxic effects of nanosilver was systematically evaluated. Our results show that nanosilver toxicity was strongly affected by the composition of the culture media. The chloride concentration, as well as the carbon content affected the silver agglomeration and the complex formation. But also the dissolution of nanosilver and the availability of free silver ions (Ag Silver coatings are used in health care products due to their bacteriostatic or antibacterial properties. The assessment of the toxicity of a certain compound is mostly done using in vitro assays. Therefore, cytotoxicity studies of nanosilver using human cell cultures have to be undertaken under well controlled and understood cultivations conditions in order to improve the compatibility of different studies. Especially when eukaryotic versus prokaryotic systems are compared for the evaluation of the use of nanosilver as antibacterial coatings for implants in order to prevent bacterial colonization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture