Merck
  • Increased toxic urinary cations in males with interstitial cystitis: a possible cause of bladder symptoms.
Increased toxic urinary cations in males with interstitial cystitis: a possible cause of bladder symptoms.
World journal of urology (2016-03-31)
Sulabha Argade, Zoltan Berecz, Yongxuan Su, C Lowell Parsons
ZUSAMMENFASSUNG
To identify and quantify toxic urinary cations in male patients with bladder pain syndrome/interstitial cystitis versus male controls, to compare them in symptomatic patients to those significantly improved, and to evaluate cytotoxicity of these cations to cultured urothelial cells to determine whether Tamm-Horsfall protein (THP) can neutralize the cations. Isolation of cationic fraction (CFs) was achieved by solid phase extraction on urine specimens of 51 male patients with IC and 33 male controls. C CF content was significantly higher in patients compared to controls (p < 0.001). Patients had higher levels of modified nucleosides, amino acids, and their derivatives compared to controls. Cytotoxicity for control versus patient mean (SEM) percent was 1.7 (2.9) % versus 63.0 (3.7) %, respectively, (p < 0.001). Cytotoxicity of metabolites was reduced in the presence of THP of control compared to THP of patient (p < 0.001). Patients with IC had significantly higher levels of cationic metabolites with higher cytotoxicity compared to controls. THP of these patients had reduced ability to sequester cytotoxicity of cationic metabolites. Patients who significantly improved on therapy had the same levels and toxicity of cationic metabolites as symptomatic males, suggesting that these cations may be the cause of epithelial dysfunction in IC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Kynurenin, ≥95.0% (NT)