Merck
CFTR Controls the Activity of NF-κB by Enhancing the Degradation of TRADD.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2016-12-14)
Hua Wang, Liudmila Cebotaru, Ha Won Lee, QingFeng Yang, Bette S Pollard, Harvey B Pollard, William B Guggino
ZUSAMMENFASSUNG
Chronic lung infection in cystic fibrosis leads to an inflammatory response that persists because of the chronic presence of bacteria and ultimately leads to a catastrophic failure of lung function. We use a combination of biochemistry, cell and molecular biology to study the interaction of TRADD, a key adaptor molecule in TNFα signaling, with CFTR in the regulation of NFκB. We show that Wt CFTR binds to and colocalizes with TRADD. TRADD is a key signaling intermediate connecting TNFα with activation of NFκB. By contrast, ΔF508 CFTR does not bind to TRADD. NF-κB activation is higher in CFBE expressing ΔF508 CFTR than in cells expressing Wt CFTR. However, this differential effect is abolished when TRADD levels are knocked down. Transfecting Wt CFTR into CFBE cells reduces NF-κB activity. However the reduction is abolished by the CFTR chloride transport inhibitor-172. Consistently, transfecting in the correctly trafficked CFTR conduction mutants G551D or S341A also fail to reduce NFκB activity. Thus CFTR must be functional if it is to regulate NF-κB activity. We also found that TNFα produced a greater increase in NF-κB activity in CFBE cells than in the same cell when Wt CFTR-corrected. Consistently, the effect is also abolished when TRADD is knocked down by shRNA. Thus, Wt CFTR control of TRADD modulates the physiological activation of NF-κB by TNFα. Based on studies with proteosomal and lysosomal inhibitors, the mechanism by which Wt CFTR, but not ΔF508 CFTR, suppresses TRADD is by lysosomal degradation. We have uncovered a novel mechanism whereby Wt CFTR regulates TNFα signaling by enhancing TRADD degradation. Thus by reducing the levels of TRADD, Wt CFTR suppresses downstream proinflammatory NFκB signaling. By contrast, suppression of NF-κB activation fails in CF cells expressing ΔF508 CFTR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
MISSION® esiRNA, targeting human TRADD