Merck
  • Prognostic significance of AKR1B10 gene expression in hepatocellular carcinoma and surrounding non-tumorous liver tissue.
Prognostic significance of AKR1B10 gene expression in hepatocellular carcinoma and surrounding non-tumorous liver tissue.
Oncology letters (2017-01-21)
Fuminori Sonohara, Yoshikuni Inokawa, Mitsuhiro Hishida, Mitsuro Kanda, Yoko Nishikawa, Suguru Yamada, Tsutomu Fujii, Hiroyuki Sugimoto, Yasuhiro Kodera, Shuji Nomoto
ZUSAMMENFASSUNG
When assessing outcome in hepatocellular carcinoma (HCC), it is important to consider prognostic factors in background non-tumorous liver tissue as well as in the tumor, since multiple occurrence is associated with background liver status such as hepatitis. The current study aimed to elucidate molecular prognostic predictors that have an association with HCC background non-tumorous tissue. Microarray expression profiling identified aldo-keto reductase family 1, member B10 (AKR1B10) as a putative non-tumorous prognostic factor, and
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%