Merck
  • Selective and vertical microfabrication of lipid tubule arrays on glass substrates using template-guided gentle hydration.
Selective and vertical microfabrication of lipid tubule arrays on glass substrates using template-guided gentle hydration.
Lab on a chip (2016-11-05)
Min Cheol Park, Pavithra Sukumar, Sang Kyung Kim, Ji Yoon Kang, Andreas Manz, Tae Song Kim
ZUSAMMENFASSUNG
Generally, asymmetric tubular lipid structures have been formed under the specific condition of gentle hydration or by using hydrodynamic and/or electrical elongation of vesicular lipid structures. Small-size lipid tubes are, however, very difficult to allocate or align in the vertical direction on the specific site of the substrate and, therefore, the ability to produce them selectively and in large quantities as an array form is limited. Herein, we propose an easy and novel method to fabricate selective and vertical lipid tube arrays using template-guided gentle hydration of dried lipid films without any external forces. A lipid solution was drop-dispensed onto a porous membrane and dried to form a lipid film. Then, the lipid-coated porous membrane was transferred to a glass substrate by using a UV-cured polymer layer to achieve tight bonding. Upon swelling with an appropriate buffer, expansion forces due to osmotic pressure during the gentle hydration process were highly constrained to confined pores, thereby resulting in the nucleation of tube-like lipid structures through the pores. Interestingly, according to the aspect ratio of pores (AR
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycero-3-Phosphocholin, lyophilized powder