- Controlled Sub-Nanometer Epitope Spacing in a Three-Dimensional Self-Assembled Peptide Hydrogel.
Controlled Sub-Nanometer Epitope Spacing in a Three-Dimensional Self-Assembled Peptide Hydrogel.
ACS nano (2016-12-28)
E Thomas Pashuck, Benoît J R Duchet, Catherine S Hansel, Stephanie A Maynard, Lesley W Chow, Molly M Stevens
PMID28024362
ZUSAMMENFASSUNG
Cells in the body use a variety of mechanisms to ensure the specificity and efficacy of signal transduction. One way that this is achieved is through tight spatial control over the position of different proteins, signaling sequences, and biomolecules within and around cells. For instance, the extracellular matrix protein fibronectin presents RGDS and PHSRN sequences that synergistically bind the α