Merck
  • Tartrate-Resistant Acid Phosphatase Deficiency in the Predisposition to Systemic Lupus Erythematosus.
Tartrate-Resistant Acid Phosphatase Deficiency in the Predisposition to Systemic Lupus Erythematosus.
Arthritis & rheumatology (Hoboken, N.J.) (2016-07-09)
Jie An, Tracy A Briggs, Audrey Dumax-Vorzet, Marta E Alarcón-Riquelme, Alexandre Belot, Michael Beresford, Ian N Bruce, Claudia Carvalho, Laurence Chaperot, Johan Frostegård, Joel Plumas, Gillian I Rice, Timothy J Vyse, Alice Wiedeman, Yanick J Crow, Keith B Elkon
ZUSAMMENFASSUNG
Mutations in the ACP5 gene, which encodes tartrate-resistant acid phosphatase (TRAP), cause the immuno-osseous disorder spondyloenchondrodysplasia, which includes as disease features systemic lupus erythematosus (SLE) and a type I interferon (IFN) signature. Our aims were to identify TRAP substrates, determine the consequences of TRAP deficiency in immune cells, and assess whether ACP5 mutations are enriched in sporadic cases of SLE. Interaction between TRAP and its binding partners was tested by a yeast 2-hybrid screening, confocal microscopy, and immunoprecipitation/Western blotting. TRAP knockdown was performed using small interfering RNA. Phosphorylation of osteopontin (OPN) was analyzed by mass spectrometry. Nucleotide sequence analysis of ACP5 was performed by Sanger sequencing or next-generation sequencing. TRAP and OPN colocalized and interacted in human macrophages and plasmacytoid dendritic cells (PDCs). TRAP dephosphorylated 3 serine residues on specific OPN peptides. TRAP knockdown resulted in increased OPN phosphorylation and increased nuclear translocation of IRF7 and P65, with resultant heightened expression of IFN-stimulated genes and IL6 and TNF following Toll-like receptor 9 stimulation. An excess of heterozygous ACP5 missense variants was observed in SLE compared to controls (P = 0.04), and transfection experiments revealed a significant reduction in TRAP activity in a number of variants. Our findings indicate that TRAP and OPN colocalize and that OPN is a substrate for TRAP in human immune cells. TRAP deficiency in PDCs leads to increased IFNα production, providing at least a partial explanation for how ACP5 mutations cause lupus in the context of spondyloenchondrodysplasia. Detection of ACP5 missense variants in a lupus cohort suggests that impaired TRAP functioning may increase susceptibility to sporadic lupus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)