Merck
  • The Effect of Superparamagnetic Iron Oxide Nanoparticle Surface Charge on Antigen Cross-Presentation.
The Effect of Superparamagnetic Iron Oxide Nanoparticle Surface Charge on Antigen Cross-Presentation.
Nanoscale research letters (2017-01-20)
Yongbin Mou, Yun Xing, Hongyan Ren, Zhihua Cui, Yu Zhang, Guangjie Yu, Walter J Urba, Qingang Hu, Hongming Hu
ZUSAMMENFASSUNG
Magnetic nanoparticles (NPs) of superparamagnetic iron oxide (SPIO) have been explored for different kinds of applications in biomedicine, mechanics, and information. Here, we explored the synthetic SPIO NPs as an adjuvant on antigen cross-presentation ability by enhancing the intracellular delivery of antigens into antigen presenting cells (APCs). Particles with different chemical modifications and surface charges were used to study the mechanism of action of antigen delivery. Specifically, two types of magnetic NPs, γFe
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-2,3-Dimercaptobernsteinsäure, ~98%