- Constituents from the leaves of Clausena lansium and their anti-inflammatory activity.
Constituents from the leaves of Clausena lansium and their anti-inflammatory activity.
Journal of natural medicines (2016-08-20)
De-Yang Shen, Ping-Chung Kuo, Shiow-Chyn Huang, Tsong-Long Hwang, Yu-Yi Chan, Po-Chuen Shieh, Nguyen Thi Ngan, Tran Dinh Thang, Tian-Shung Wu
PMID27539584
ZUSAMMENFASSUNG
Five new acyclic amides, clausenalansamides C-G (1-5), clausenaline G (6) and (±)-5-(4-methylphenyl)-γ-valerolactone (7) reported from the natural source for the first time, were characterized from the leaves of Clausena lansium. Their structures were established by spectroscopic and spectrometric methods, and the absolute configurations of new compounds 1-5 were determined by electronic circular dichroism and single-crystal X-ray diffraction analyses. Eighteen compounds were evaluated for their anti-inflammatory activity and only imperatorin (11) and wampetin (12) displayed significant inhibition of fMLP/CB-induced superoxide anion generation with IC