Merck
  • Silencing of Armadillo Repeat-Containing Protein 8 (ARMc8) Inhibits TGF-β-Induced EMT in Bladder Carcinoma UMUC3 Cells.
Silencing of Armadillo Repeat-Containing Protein 8 (ARMc8) Inhibits TGF-β-Induced EMT in Bladder Carcinoma UMUC3 Cells.
Oncology research (2017-01-14)
Xuan Liang, Qun-Li Men, Yong-Wei Li, He-Cheng Li, Tie Chong, Zhao-Lun Li
ZUSAMMENFASSUNG
Armadillo repeat-containing protein 8 (ARMc8) is a key factor in regulating cell migration, proliferation, tissue maintenance, and tumorigenesis. However, its role in bladder cancer remains unknown. Thus, in this study we sought to investigate the effect of ARMc8 on the epithelial-to-mesenchymal transition (EMT) progress in bladder cancer cells induced by transforming growth factor-β1 (TGF-β1). Our results found that ARMc8 was highly expressed in bladder cancer cell lines. ARMc8 silencing inhibited the TGF-β1-induced migration and invasion and suppressed the EMT progress in bladder cancer cells. Furthermore, ARMc8 silencing inhibited the TGF-β1-induced expression of β-catenin, cyclin D1, and c-myc in bladder cancer cells. In conclusion, the present study demonstrates a novel function for ARMc8, which acts as a mediator for TGF-β1-induced cell migration/invasion through modulation of the Wnt/β-catenin signaling pathway in bladder cancer cells. This study suggests that ARMc8 may be a potential therapeutic target for the development of therapies for bladder cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human GPR182