Merck
  • Accessory pathway location affects brain natriuretic peptide level in patients with Wolff-Parkinson-White syndrome.
Accessory pathway location affects brain natriuretic peptide level in patients with Wolff-Parkinson-White syndrome.
Journal of interventional cardiac electrophysiology : an international journal of arrhythmias and pacing (2016-11-07)
Yosuke Nakatani, Koji Kumagai, Shigeto Naito, Kohki Nakamura, Kentaro Minami, Masahiro Nakano, Takehito Sasaki, Koichiro Kinugawa, Shigeru Oshima
ZUSAMMENFASSUNG
The purpose of this study was to investigate the relationship between the accessory pathway location and brain natriuretic peptide (BNP) level in patients with Wolff-Parkinson-White (WPW) syndrome. We divided 102 WPW syndrome patients with normal left ventricular systolic function into four groups: those with manifest right (MR, n = 14), manifest septal (MS, n = 11), manifest left (ML, n = 30), and concealed (C, n = 47) accessory pathways. BNP level and electrophysiological properties, including difference in timing of the ventricular electrogram between the His bundle area and the distal coronary sinus area (His-CS delay), which indicate intraventricular dyssynchrony, were compared. BNP levels (pg/dl) were higher in the MR and MS groups than in the ML and C groups (MR, 64 ± 58; MS, 55 ± 45; ML, 17 ± 15; C, 25 ± 21; P < 0.001). AV intervals (ms) were shorter in the MR and MS groups than in the ML and C groups (MR, 76 ± 16; MS, 83 ± 6; ML, 101 ± 19; C, 136 ± 20; P < 0.001). His-CS delay (ms) was longer in the MR group than in the other groups (MR, 50 ± 15; MS, 21 ± 7; ML, 23 ± 10; C, 19 ± 8; P < 0.001). The AV interval (P < 0.01) and the His-CS delay (P < 0.001) were negatively and positively correlated, respectively, with the BNP level. Anterograde conduction with a right or septal accessory pathway increased the BNP level in WPW syndrome patients with normal cardiac function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(+)-Biotin-4-nitrophenylester, 98%