Merck
Capture of 2D Microparticle Arrays via a UV-Triggered Thiol-yne "Click" Reaction.
Advanced materials (Deerfield Beach, Fla.) (2016-10-08)
Debora Walker, Dhruv P Singh, Peer Fischer
ZUSAMMENFASSUNG
Immobilization of colloidal assemblies onto solid supports via a fast UV-triggered click-reaction is achieved. Transient assemblies of microparticles and colloidal materials can be captured and transferred to solid supports. The technique does not require complex reaction conditions, and is compatible with a variety of particle assembly methods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%