Merck
  • Hypoxia acclimation and subsequent reoxygenation partially prevent Mn-induced damage in silver catfish.
Hypoxia acclimation and subsequent reoxygenation partially prevent Mn-induced damage in silver catfish.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2016-10-28)
G S Dolci, H Z Rosa, R C S Barcelos, L T Vey, A Santos, P DallaVechia, C Bizzi, M A Cunha, B Baldisserotto, M E Burger
ZUSAMMENFASSUNG
This study investigated if hypoxia acclimation modifies the hematological and oxidative profiles in tissues of Mn-exposed silver catfish (Rhamdia quelen), and if such modifications persist upon subsequent reoxygenation. Silver catfish acclimated to hypoxia (~3mgL
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Ouabain, European Pharmacopoeia (EP) Reference Standard