Merck
  • Relationship between IL-10 gene -819C/T polymorphism and the risk of inflammatory bowel disease: a meta-analysis
Relationship between IL-10 gene -819C/T polymorphism and the risk of inflammatory bowel disease: a meta-analysis
African Health Sciences (2016)
Haien Wu