Merck
  • Microparticles mediate MRP1 intercellular transfer and the re-templating of intrinsic resistance pathways.
Microparticles mediate MRP1 intercellular transfer and the re-templating of intrinsic resistance pathways.
Pharmacological Research (2013)
Lu JF et al.