Merck
  • Angiotensin II cell signaling: physiological and pathological effects in the cardiovascular system.
Angiotensin II cell signaling: physiological and pathological effects in the cardiovascular system.
American Journal of Physiology. Cell Physiology (2007)
Mehta PK and Griendling KK